2-Trifluoromethyl-quinazolin-4-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Trifluoromethyl)quinazolin-4(3H)-one

059274-500MG 059274-1G 059274-5G
101917-746EA 466.15 USD
101917-746 101917-748 101917-750
2-Trifluoromethyl-quinazolin-4-ol ≥95%
2-Trifluoromethyl-quinazolin-4-ol

Hazard: Irritant

Formula: C₉H₅F₃N₂O
MW: 214.15 g/mol
MDL Number: MFCD01764613
CAS Number: 26059-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR