3-(4-Trifluoromethylpyridin-2-yl)benzoic acid methyl ester ≥95%

Supplier: Matrix Scientific

059271-1G 059271-5G
101917-734EA 553.85 USD
101917-734 101917-736
3-(4-Trifluoromethylpyridin-2-yl)benzoic acid methyl ester ≥95%
3-(4-Trifluoromethylpyridin-2-yl)benzoic acid methyl ester

Hazard: Irritant

Formula: C₁₄H₁₀F₃NO₂
MDL Number: MFCD18384863
CAS Number: 1299607-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR