5-(Trifluoromethyl)pyridin-2-ylamidoxime ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Trifluoromethyl)pyridine-2-carboxamide oxime

059266-1G 059266-5G
101917-720EA 727.69 USD
101917-720 101917-722
5-(Trifluoromethyl)pyridin-2-ylamidoxime ≥95%
5-(Trifluoromethyl)pyridin-2-ylamidoxime

Hazard: Irritant

Formula: C₇H₆F₃N₃O
MDL Number: MFCD00205809
CAS Number: 175277-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR