3-(Trifluoromethoxy)benzamidoxime ≥95%

Supplier: Matrix Scientific

059262-1G 059262-5G
101917-704EA 233.85 USD
101917-704 101917-706
3-(Trifluoromethoxy)benzamidoxime ≥95%
3-(Trifluoromethoxy)benzamidoxime

Hazard: Irritant

Formula: C₈H₇F₃N₂O₂
MDL Number: MFCD06660166
CAS Number: 886500-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR