2-(2-Thienyl)pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Thiophen-2-yl)pyridine, 2-(2-Pyridyl)thiophene

059259-1G 059259-5G
101917-688EA 104.62 USD
101917-688 101917-690
2-(2-Thienyl)pyridine ≥95%
2-(2-Thienyl)pyridine

Hazard: Irritant

Formula: C₉H₇NS
MW: 161.22 g/mol
MDL Number: MFCD00044441
CAS Number: 3319-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR