Thiophene-3-methylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Thienylmethylamine

059258-1G 059258-5G
101917-684EA 495.38 USD
101917-684 101917-686
Thiophene-3-methylamine ≥95%
Thiophene-3-methylamine

Hazard: Corrosive

Formula: C₅H₇NS
MDL Number: MFCD01529872
CAS Number: 27757-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR