Thiophene-2-carboxamidine hydrochloride ≥95%

Supplier: Matrix Scientific

059250-1G 059250-10G
101917-658EA 153.85 USD
101917-658 101917-660
Thiophene-2-carboxamidine hydrochloride ≥95%
Thiophene-2-carboxamidine hydrochloride

Hazard: Irritant

Formula: C₅H₇ClN₂S
Melting Pt: 173…177 °C
MDL Number: MFCD00173786
CAS Number: 54610-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR