Quinoxaline-2-carbonyl chloride ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Quinoxaloyl chloride
059243-250MG
059243-1G
101917-638EA
132.31
USD
101917-638
101917-640
Quinoxaline-2-carbonyl chloride ≥95%
Quinoxaline-2-carbonyl chloride
Hazard: Irritant
Formula:
C₉H₅ClN₂O MW: 192.61 g/mol |
MDL Number:
MFCD00006725 CAS Number: 54745-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...