2-(3-Pyridinyl)-4-thiazolecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Pyridin-3-yl)thiazole-4-carboxylic acid

059239-1G 059239-5G 059239-10G
101917-620EA 378.46 USD
101917-620 101917-622 101917-624
2-(3-Pyridinyl)-4-thiazolecarboxylic acid ≥95%
2-(3-Pyridinyl)-4-thiazolecarboxylic acid

Hazard: Irritant

Formula: C₉H₆N₂O₂S
MW: 206.22 g/mol
MDL Number: MFCD00052304
CAS Number: 39067-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR