3-Pyridin-2-yl[1,2,4]oxadiazole-5-carboxylic acid ethyl ester ≥95%

Supplier: Matrix Scientific

059238-500MG 059238-1G 059238-5G
101917-614EA 466.15 USD
101917-614 101917-616 101917-618
3-Pyridin-2-yl[1,2,4]oxadiazole-5-carboxylic acid ethyl ester ≥95%
3-Pyridin-2-yl[1,2,4]oxadiazole-5-carboxylic acid ethyl ester

Hazard: Irritant

Formula: C₁₀H₉N₃O₃
MDL Number: MFCD09908124
CAS Number: 163719-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR