3,4-Pyridinedicarboximide ≥95%

Supplier: Matrix Scientific

059237-1G 059237-5G 059237-25G
101917-608EA 49.23 USD
101917-608 101917-610 102833-276
3,4-Pyridinedicarboximide ≥95%
3,4-Pyridinedicarboximide

Hazard: Irritant

Formula: C₇H₄N₂O₂
Melting Pt: 230…232 °C
MDL Number: MFCD00013439
CAS Number: 4664-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR