1-(2H-Pyrazol-3-yl)ethanone hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Acetyl-1H-pyrazole hydrochloride, 1-(1H-Pyrazol-3-yl)ethanone hydrochloride

059235-1G 059235-5G
101917-600EA 195.38 USD
101917-600 101917-602
1-(2H-Pyrazol-3-yl)ethanone hydrochloride ≥95%
1-(2H-Pyrazol-3-yl)ethanone hydrochloride

Hazard: Irritant

Formula: C₅H₇ClN₂O
MW: 146.58 g/mol
MDL Number: MFCD00204160
CAS Number: 175277-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR