4-(6-Piperazin-1-yl-4-trifluoromethylpyridin-2-yl)morpholine ≥95%

Supplier: Matrix Scientific

059230-1G 059230-5G
101917-582EA 313.85 USD
101917-582 101917-584
4-(6-Piperazin-1-yl-4-trifluoromethylpyridin-2-yl)morpholine ≥95%
4-(6-Piperazin-1-yl-4-trifluoromethylpyridin-2-yl)morpholine

Hazard: Irritant

Formula: C₁₄H₁₉F₃N₄O
MDL Number: MFCD11052383
CAS Number: 1089330-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR