4-Phenyl[1,3,5]triazine-2-thiol ≥95%

Supplier: Matrix Scientific

059227-1G 059227-5G
101917-570EA 436.92 USD
101917-570 101917-572
4-Phenyl[1,3,5]triazine-2-thiol ≥95%
4-Phenyl[1,3,5]triazine-2-thiol

Hazard: Irritant

Formula: C₉H₇N₃S
MDL Number: MFCD09702439
CAS Number: 1000018-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR