2-Phenylthioacetamide ≥95%

Supplier: Matrix Scientific

059225-1G 059225-5G
101917-560EA 161.54 USD
101917-560 101917-562
2-Phenylthioacetamide ≥95%
2-Phenylthioacetamide

Hazard: Irritant

Formula: C₈H₉NS
Melting Pt: 91…95 °C
MDL Number: MFCD00022177
CAS Number: 645-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR