1-(3-Phenyl-[1,2,4]-thiadiazol-5-yl)piperazine ≥95%

Supplier: Matrix Scientific

059222-500MG 059222-1G 059222-5G
101917-546EA 175.38 USD
101917-546 101917-548 101917-550
1-(3-Phenyl-[1,2,4]-thiadiazol-5-yl)piperazine ≥95%
1-(3-Phenyl-[1,2,4]-thiadiazol-5-yl)piperazine

Hazard: Irritant

Formula: C₁₂H₁₄N₄S
MW: 246.34 g/mol
MDL Number: MFCD00115115
CAS Number: 306935-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR