6-Phenylpyrimidine-2,4-diol ≥95%

Supplier: Matrix Scientific

059221-500MG 059221-1G 059221-5G
101917-540EA 310.77 USD
101917-540 101917-542 101917-544
6-Phenylpyrimidine-2,4-diol ≥95%
6-Phenylpyrimidine-2,4-diol

Hazard: Irritant

Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
MDL Number: MFCD00128171
CAS Number: 13345-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR