5-Phenyl[1,2,4]oxadiazole-3-carboxylic acid ethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-phenyl-1,2,4-oxadiazole-3-carboxylate

059217-1G 059217-5G 059217-10G
101917-526EA 215.38 USD
101917-526 101917-528 101917-530
5-Phenyl[1,2,4]oxadiazole-3-carboxylic acid ethyl ester ≥95%
5-Phenyl[1,2,4]oxadiazole-3-carboxylic acid ethyl ester

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₃
MW: 218.21 g/mol
MDL Number: MFCD01764002
CAS Number: 37384-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR