N'-[2-Phenoxy-1-(2,2,2-trifluoroethylamino)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

059210-500MG 059210-1G
101917-502EA 284.62 USD
101917-502 101917-504
N'-[2-Phenoxy-1-(2,2,2-trifluoroethylamino)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[2-Phenoxy-1-(2,2,2-trifluoroethylamino)ethylidene]hydrazinecarboxylic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₅H₂₀F₃N₃O₃
MW: 347.34 g/mol
MDL Number: MFCD10568260
CAS Number: 1053657-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR