4-Oxotetrahydrothiophene-3-carboxylic acid methyl ester, Technical Grade

Supplier: Matrix Scientific

Synonyms: Methyl 4-oxotetrahydrothiophene-3-carboxylate

059208-1G 059208-5G
101917-492EA 190.77 USD
101917-492 101917-494
4-Oxotetrahydrothiophene-3-carboxylic acid methyl ester, Technical Grade
4-Oxotetrahydrothiophene-3-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₆H₈O₃S
MW: 160.19 g/mol
MDL Number: MFCD00052381
CAS Number: 2689-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR