3-Oxopyrrolidine-1,2-dicarboxylic acid-1-tert-butyl ester-2-ethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl N-Boc-3-oxopyrrolidine-2-carboxylate, 1-tert-Butyl 2-ethyl 3-oxopyrrolidine-1,2-dicarboxylate

059207-1G 059207-5G
101917-488EA 306.15 USD
101917-488 101917-490
3-Oxopyrrolidine-1,2-dicarboxylic acid-1-tert-butyl ester-2-ethyl ester ≥95%
3-Oxopyrrolidine-1,2-dicarboxylic acid-1-tert-butyl ester-2-ethyl ester

Hazard: Irritant

Formula: C₁₂H₁₉NO₅
MW: 257.29 g/mol
MDL Number: MFCD11052440
CAS Number: 170123-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR