2-Nitro-4-thiophenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Nitrothiophene-3-carboxylic acid

059199-1G 059199-5G
101917-458EA 89.23 USD
101917-458 101917-460
2-Nitro-4-thiophenecarboxylic acid ≥95%
2-Nitro-4-thiophenecarboxylic acid

Hazard: Irritant

Formula: C₅H₃NO₄S
MDL Number: MFCD00052582
CAS Number: 40357-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR