3-Morpholin-4-yl-5-trifluoromethyl-benzonitrile ≥95%
Supplier: Matrix Scientific
059191-1G
059191-5G
101917-430EA
436.92
USD
101917-430
101917-432
3-Morpholin-4-yl-5-trifluoromethyl-benzonitrile ≥95%
3-Morpholin-4-yl-5-trifluoromethyl-benzonitrile
Hazard: Irritant
Formula:
C₁₂H₁₁F₃N₂O |
MDL Number:
MFCD11845701 CAS Number: 220954-14-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...