3-Morpholin-4-yl-5-trifluoromethylbenzoic acid ≥95%

Supplier: Matrix Scientific

059190-1G 059190-5G
101917-426EA 495.38 USD
101917-426 101917-428
3-Morpholin-4-yl-5-trifluoromethylbenzoic acid ≥95%
3-Morpholin-4-yl-5-trifluoromethylbenzoic acid

Hazard: Irritant

Formula: C₁₂H₁₂F₃NO₃
MDL Number: MFCD11845699
CAS Number: 250682-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR