2-(Morpholin-4'-yl)phenylacetamide hydrochloride ≥95%

Supplier: Matrix Scientific

059188-1G 059188-5G
101917-420EA 553.85 USD
101917-420 101917-422
2-(Morpholin-4'-yl)phenylacetamide hydrochloride ≥95%
2-(Morpholin-4'-yl)phenylacetamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₂
MDL Number: MFCD18384860
CAS Number: 1299607-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR