3-(4-Morpholinyl)aniline ≥95%

Supplier: Matrix Scientific

059187-1G 059187-5G
101917-416EA 32.31 USD
101917-416 101917-418
3-(4-Morpholinyl)aniline ≥95%
3-(4-Morpholinyl)aniline

Hazard: Irritant

Formula: C10H14N2O
MW: 178.2300 g/mol
MDL Number: MFCD03197165
CAS Number: 159724-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR