2-Methylthiazole-4-carboxamidine hydrochloride ≥95%

Supplier: Matrix Scientific

059177-1G 059177-5G 059177-10G
101917-384EA 307.69 USD
101917-384 101917-386 101917-388
2-Methylthiazole-4-carboxamidine hydrochloride ≥95%
2-Methylthiazole-4-carboxamidine hydrochloride

Hazard: Irritant

Formula: C₅H₈ClN₃S
MW: 177.66 g/mol
MDL Number: MFCD00178765
CAS Number: 18876-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR