(3S,4R)-1-Methylpyrrolidine-3,4-dicarboxylic acid dimethyl ester racemate ≥95%

Supplier: Matrix Scientific

059175-1G 059175-5G
101917-378EA 553.85 USD
101917-378 101917-380
(3S,4R)-1-Methylpyrrolidine-3,4-dicarboxylic acid dimethyl ester racemate ≥95%
(3S,4R)-1-Methylpyrrolidine-3,4-dicarboxylic acid dimethyl ester

Hazard: Irritant

Formula: C₉H₁₅NO₄
MDL Number: MFCD11845731
CAS Number: 102389-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR