1-Methylpyrrolidine-3-carboxylicacid tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 1-methylpyrrolidine-3-carboxylate

059174-1G 059174-5G
101917-374EA 495.38 USD
101917-374 101917-376
1-Methylpyrrolidine-3-carboxylicacid tert-butyl ester ≥95%
1-Methylpyrrolidine-3-carboxylicacid tert-butyl ester

Hazard: Irritant

Formula: C₁₀H₁₉NO₂
MDL Number: MFCD11227161
CAS Number: 635700-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR