6-Methylpyridine-2-carboxamidine hydrochloride ≥95%

Supplier: Matrix Scientific

059172-1G 059172-5G
101917-368EA 1372.31 USD
101917-368 101917-370
6-Methylpyridine-2-carboxamidine hydrochloride ≥95%
6-Methylpyridine-2-carboxamidine hydrochloride

Hazard: Irritant

Formula: C₇H₉N₃·ClH
MW: 171.627 g/mol
MDL Number: MFCD09952702
CAS Number: 112736-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR