1-Methyl-4-piperidin-4-ylpiperazine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Methyl-4-(piperidin-4-yl)piperazine

059170-5G 059170-10G
101917-360EA 161.54 USD
101917-360 101917-362
1-Methyl-4-piperidin-4-ylpiperazine ≥95%
1-Methyl-4-piperidin-4-ylpiperazine

Hazard: Irritant

Formula: C₁₀H₂₁N₃
MW: 183.3 g/mol
MDL Number: MFCD03274729
CAS Number: 53617-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR