(4-Methylpiperazin-1-yl)phenylacetonitrile hydrochloride ≥95%

Supplier: Matrix Scientific

059169-1G 059169-5G 059169-10G
101917-354EA 204.62 USD
101917-354 101917-356 101917-358
(4-Methylpiperazin-1-yl)phenylacetonitrile hydrochloride ≥95%
(4-Methylpiperazin-1-yl)phenylacetonitrile hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClN₃
MW: 251.76 g/mol
MDL Number: MFCD11052463
CAS Number: 108953-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR