[3-Methyl-5-phenyl-4-isoxazolyl]methanol ≥95%
Supplier: Matrix Scientific
059165-1G
059165-5G
059165-10G
101917-342EA
150.77
USD
101917-342
101917-344
101917-346
[3-Methyl-5-phenyl-4-isoxazolyl]methanol ≥95%
[3-Methyl-5-phenyl-4-isoxazolyl]methanol
Hazard: Irritant
Formula:
C₁₁H₁₁NO₂ |
CAS Number:
113826-87-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...