3-Methyl-5-phenylisoxazole-4-carboxylic acid methyl ester ≥95%

Supplier: Matrix Scientific

059164-1G 059164-5G
101917-338EA 553.85 USD
101917-338 101917-340
3-Methyl-5-phenylisoxazole-4-carboxylic acid methyl ester ≥95%
3-Methyl-5-phenylisoxazole-4-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₁₂H₁₁NO₃
MDL Number: MFCD06204160
CAS Number: 773876-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR