5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid ethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-methyl-1-phenyl-1H-pyrazole-4-carboxylate

059162-1G 059162-5G 059162-10G
101917-330EA 204.62 USD
101917-330 101917-332 101917-334
5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid ethyl ester ≥95%
5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid ethyl ester

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₂
MW: 230.27 g/mol
MDL Number: MFCD00020741
CAS Number: 89193-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR