4-Methyl-2-[(1-phenylcyclopentanecarbonyl)amino]thiazole-5-carboxylic acid ethyl ester ≥95%

Supplier: Matrix Scientific

059161-500MG 059161-1G
101917-326EA 553.85 USD
101917-326 101917-328
4-Methyl-2-[(1-phenylcyclopentanecarbonyl)amino]thiazole-5-carboxylic acid ethyl ester ≥95%
4-Methyl-2-[(1-phenylcyclopentanecarbonyl)amino]thiazole-5-carboxylic acid ethyl ester

Hazard: Irritant

Formula: C₁₉H₂₂N₂O₃S
MW: 358.46 g/mol
MDL Number: MFCD03839529
CAS Number: 1024162-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR