3-Methyl[1,2,4]oxadiazole-5-carboxylic acid ethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-methyl-1,2,4-oxadiazole-5-carboxylate

059158-1G 059158-5G 059158-10G
101917-310EA 349.23 USD
101917-310 101917-312 101917-314
3-Methyl[1,2,4]oxadiazole-5-carboxylic acid ethyl ester ≥95%
3-Methyl[1,2,4]oxadiazole-5-carboxylic acid ethyl ester

Hazard: Irritant

Formula: C₆H₈N₂O₃
MW: 156.14 g/mol
MDL Number: MFCD08275042
CAS Number: 40019-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR