5-Methyl-4-nitro-3-trifluoromethyl-1H-pyrazole ≥95%

Supplier: Matrix Scientific

059157-1G 059157-5G 059157-10G
101917-304EA 115.38 USD
101917-304 101917-306 101917-308
5-Methyl-4-nitro-3-trifluoromethyl-1H-pyrazole ≥95%
5-Methyl-4-nitro-3-trifluoromethyl-1H-pyrazole

Hazard: Irritant

Formula: C₅H₄F₃N₃O₂
MW: 195.1 g/mol
Melting Pt: 100 °C
MDL Number: MFCD00156055
CAS Number: 27116-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR