2-Methyl-4-(3-nitro-phenyl)thiazole hydrobromide ≥95%

Supplier: Matrix Scientific

059155-1G 059155-5G 059155-10G
101917-296EA 320 USD
101917-296 101917-298 101917-300
2-Methyl-4-(3-nitro-phenyl)thiazole hydrobromide ≥95%
2-Methyl-4-(3-nitro-phenyl)thiazole hydrobromide

Hazard: Irritant

Formula: C₁₀H₉BrN₂O₂S
MDL Number: MFCD08832496
CAS Number: 1055195-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR