5-Methyl-4-nitro-2H-pyrazole-3-carboxylic acid ethyl ester ≥95%

Supplier: Matrix Scientific

059153-1G 059153-5G
101917-290EA 553.85 USD
101917-290 101917-292
5-Methyl-4-nitro-2H-pyrazole-3-carboxylic acid ethyl ester ≥95%
5-Methyl-4-nitro-2H-pyrazole-3-carboxylic acid ethyl ester

Hazard: Irritant

Formula: C₇H₉N₃O₄
MW: 199.17 g/mol
MDL Number: MFCD02947463
CAS Number: 28668-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR