5-Methyl-4-nitro-2H-pyrazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Methyl-4-nitro-1H-pyrazole-5-carboxylic acid

059152-1G 059152-5G
101917-286EA 553.85 USD
101917-286 101917-288
5-Methyl-4-nitro-2H-pyrazole-3-carboxylic acid ≥95%
5-Methyl-4-nitro-2H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₅H₅N₃O₄
MW: 171.11 g/mol
MDL Number: MFCD00458002
CAS Number: 5334-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR