3-(Hydroxymethyl)-5-methylisoxazole ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxymethyl-5-methylisoxazole, 5-Methyl-3-isoxazolemethanol

059151-1G 059151-5G
101917-282EA 112.31 USD
101917-282 101917-284
3-(Hydroxymethyl)-5-methylisoxazole ≥95%
3-(Hydroxymethyl)-5-methylisoxazole

Hazard: Irritant

Formula: C₅H₇NO₂
MW: 113.11 g/mol
MDL Number: MFCD00085129
CAS Number: 35166-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR