3-Methylisothiazol-5-ylamine hydrobromide ≥95%
Supplier: Matrix Scientific
059150-1G
059150-5G
059150-10G
101917-276EA
156.92
USD
101917-276
101917-278
101917-280
3-Methylisothiazol-5-ylamine hydrobromide ≥95%
3-Methylisothiazol-5-ylamine hydrobromide
Hazard: Irritant
Formula:
C₄H₇BrN₂S MW: 195.09 g/mol |
MDL Number:
MFCD12131107 CAS Number: 1208081-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...