3-Methylisothiazole-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

059149-500MG 059149-1G
101917-272EA 1076.92 USD
101917-272 101917-274
3-Methylisothiazole-5-carboxylic acid ≥95%
3-Methylisothiazole-5-carboxylic acid

Hazard: Irritant

Formula: C₅H₅NO₂S
MDL Number: MFCD11110341
CAS Number: 66975-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR