2-(1-Methyl-1H-pyrazol-3-yl)phenylamine ≥95%

Supplier: Matrix Scientific

059143-500MG 059143-1G
101917-252EA 553.85 USD
101917-252 101917-254
2-(1-Methyl-1H-pyrazol-3-yl)phenylamine ≥95%
2-(1-Methyl-1H-pyrazol-3-yl)phenylamine

Hazard: Irritant

Formula: C₁₀H₁₁N₃
MW: 173.22 g/mol
MDL Number: MFCD11052424
CAS Number: 87488-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR