4-(1-Methyl-1H-pyrazol-3-yl)phenylamine ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(1-Methyl-1H-pyrazol-3-yl)aniline
059141-1G
059141-5G
101917-246EA
553.85
USD
101917-246
101917-248
4-(1-Methyl-1H-pyrazol-3-yl)phenylamine ≥95%
4-(1-Methyl-1H-pyrazol-3-yl)phenylamine
Hazard: Irritant
Formula:
C₁₀H₁₁N₃ MW: 173.22 g/mol |
MDL Number:
MFCD03407387 CAS Number: 916766-82-0 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...