4-(1-Methyl-1H-pyrazol-3-yl)benzenesulfonylchloride ≥95%

Supplier: Matrix Scientific

059138-1G 059138-5G
101917-236EA 393.85 USD
101917-236 101917-238
4-(1-Methyl-1H-pyrazol-3-yl)benzenesulfonylchloride ≥95%
4-(1-Methyl-1H-pyrazol-3-yl)benzenesulfonylchloride

Hazard: Irritant

Formula: C₁₀H₉ClN₂O₂S
MW: 256.71 g/mol
MDL Number: MFCD09817463
CAS Number: 916766-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR