1-Methyl-1H-pyrazole-3,5-dicarboxylic acid dimethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Dimethyl 1-methyl-1H-pyrazole-3,5-dicarboxylate

059137-1G 059137-5G
101917-232EA 495.38 USD
101917-232 101917-234
1-Methyl-1H-pyrazole-3,5-dicarboxylic acid dimethyl ester ≥95%
1-Methyl-1H-pyrazole-3,5-dicarboxylic acid dimethyl ester

Hazard: Irritant

Formula: C₈H₁₀N₂O₄
MW: 198.18 g/mol
MDL Number: MFCD08443806
CAS Number: 33146-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR