Ethyl 5-methylpyrazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Methylpyrazole-3-carboxylic acid ethyl ester, Ethyl 5-methyl-1H-pyrazole-3-carboxylate

059135-1G 059135-5G
101917-228EA 93.85 USD
101917-228 101917-230
Ethyl 5-methylpyrazole-3-carboxylate ≥95%
Ethyl 5-methylpyrazole-3-carboxylate

Hazard: Irritant

Formula: C₇H₁₀N₂O₂
MW: 154.17 g/mol
MDL Number: MFCD00233455
CAS Number: 4027-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR