4-Methyl-1H-indole-2-carboxylic acid methyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-Methylindole-2-carboxylate

059134-500MG 059134-1G 059134-5G
101917-222EA 466.15 USD
101917-222 101917-224 101917-226
4-Methyl-1H-indole-2-carboxylic acid methyl ester ≥95%
4-Methyl-1H-indole-2-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₁₁H₁₁NO₂
MW: 189.22 g/mol
MDL Number: MFCD04966966
CAS Number: 136831-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR